Tisk…

1-Butyl-2-methylindole 95%

Dodavatel: Apollo Scientific

OR913042-5G
APOSOR913042-5GEA 3060 CZK
APOSOR913042-5G
1-Butyl-2-methylindole 95%
1-Butyl-2-methylindole
Vzorec: C₁₃H₁₇N
MW: 187,281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972048
CAS číslo: 42951-35-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR