3-(Aminoethyl)benzo[b]furan 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-(1-Benzofuran-3-yl)ethylamine, 2-(Benzo[b]furan-3-yl)ethylamine, 2-(Benzofuran-3-yl)ethanamine, 3-(Aminoethyl)-1-benzofuran
OR9130-1G
OR9130-250MG
APOSOR9130-1GEA
5260
CZK
APOSOR9130-1G
APOSOR9130-250MG
3-(Aminoethyl)benzo[b]furan 95%
3-(Aminoethyl)benzo[b]furan
Vzorec:
C₁₀H₁₁NO MW: 161,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08271905 CAS číslo: 27404-31-5 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...