Tisk…

4-Mercaptobenzyl alcohol 95%

Dodavatel: Apollo Scientific

Synonyms: (4-Mercaptophenyl)methanol

OR912890-1G OR912890-250MG
APOSOR912890-1GEA 10550 CZK
APOSOR912890-1G APOSOR912890-250MG
4-Mercaptobenzyl alcohol 95%
4-Mercaptobenzyl alcohol
Vzorec: C₇H₈OS
MW: 140,203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041529
CAS číslo: 53339-53-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR