Tisk…

7-(Aminomethyl)-2,3-dihydro-2,2-dimethylbenzo[b]furan 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 7-(Aminomethyl)-2,3-dihydro-2,2-dimethyl-1-benzofuran, (2,3-Dihydro-2,2-dimethyl-1-benzofuran-7-yl)methylamine

OR9128-1G OR9128-250MG
APOSOR9128-1GEA 16900 CZK
APOSOR9128-1G APOSOR9128-250MG
7-(Aminomethyl)-2,3-dihydro-2,2-dimethylbenzo[b]furan 97%
7-(Aminomethyl)-2,3-dihydro-2,2-dimethylbenzo[b]furan
Vzorec: C₁₁H₁₅NO
MW: 177,243 g/mol
Bod varu: 70…75 °C (0,08 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08271885
CAS číslo: 868755-44-6
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR