5-Bromo-2-methoxy-3-methylaniline 98%
Dodavatel: Apollo Scientific
OR912694-1G
OR912694-5G
APOSOR912694-1GEA
7080
CZK
APOSOR912694-1G
APOSOR912694-5G
5-Bromo-2-methoxy-3-methylaniline 98%
5-Bromo-2-methoxy-3-methylaniline
Vzorec:
C₈H₁₀BrNO MW: 216.075 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22375004 CAS číslo: 1381944-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...