Tisk…

(2-Cyanophenyl)acetic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 2-(2-Cyanophenyl)acetic acid

OR912691-5G
APOSOR912691-5GEA 1740 CZK
APOSOR912691-5G
(2-Cyanophenyl)acetic acid 98%
(2-Cyanophenyl)acetic acid
Vzorec: C₉H₇NO₂
MW: 161,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01646238
CAS číslo: 18698-99-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR