(S)-5-(Bromomethyl)-2-pyrrolidinone 96%
Dodavatel: Apollo Scientific
Synonyms:
(S)-5-Bromomethyl-2-pyrrolidinone
OR912689-5G
APOSOR912689-5GEA
5230
CZK
APOSOR912689-5G
(S)-5-(Bromomethyl)-2-pyrrolidinone 96%
(S)-5-(Bromomethyl)-2-pyrrolidinone
Vzorec:
C₅H₈BrNO MW: 178,027 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01632456 CAS číslo: 72479-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...