Tisk…

N-Methyl-4-tyramine 96%

Dodavatel: Apollo Scientific

OR912594-1G OR912594-5G
APOSOR912594-1GEA 3740 CZK
APOSOR912594-1G APOSOR912594-5G
N-Methyl-4-tyramine 96%
N-Methyl-4-tyramine
Vzorec: C₉H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00870493
CAS číslo: 370-98-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR