Ethyl 2-hydroxycyclohexanecarboxylate 95%
Dodavatel: Apollo Scientific
OR912585-1G
OR912585-250MG
APOSOR912585-1GEA
4630
CZK
APOSOR912585-1G
APOSOR912585-250MG
Ethyl 2-hydroxycyclohexanecarboxylate 95%
Ethyl 2-hydroxycyclohexanecarboxylate
Vzorec:
C₉H₁₆O₃ MW: 172,222 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00832060 CAS číslo: 3444-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...