Tisk…

N-Ethyl-3-nitrobenzamide 98%

Dodavatel: Apollo Scientific

OR912583-1G OR912583-5G
APOSOR912583-1GEA 4770 CZK
APOSOR912583-1G APOSOR912583-5G
N-Ethyl-3-nitrobenzamide 98%
N-Ethyl-3-nitrobenzamide
Vzorec: C₉H₁₀N₂O₃
MW: 194,187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01211875
CAS číslo: 26819-11-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR