4-Hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester 95%
Dodavatel: Apollo Scientific
OR912465-100G
OR912465-25G
APOSOR912465-100GEA
27080
CZK
APOSOR912465-100G
APOSOR912465-25G
4-Hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester 95%
4-Hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester
Vzorec:
C₈H₁₀N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00223606 CAS číslo: 67383-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...