Indan-5-carbaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR912348-1G
OR912348-250MG
APOSOR912348-1GEA
7340
CZK
APOSOR912348-1G
APOSOR912348-250MG
Indan-5-carbaldehyde 95%
Indan-5-carbaldehyde
Vzorec:
C₁₀H₁₀O Storage Temperature: Ambient |
MDL Number:
MFCD00086208 CAS číslo: 30084-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...