Tisk…

1-Boc-3-(N-methoxy-N-methylcarbamoyl)piperidine 97%

Dodavatel: Apollo Scientific

Synonyms: 1-Boc-N-methoxy-N-methylpiperidine-3-carboxamide

OR912064-5G
APOSOR912064-5GEA 3380 CZK
APOSOR912064-5G
1-Boc-3-(N-methoxy-N-methylcarbamoyl)piperidine 97%
1-Boc-3-(N-methoxy-N-methylcarbamoyl)piperidine
Vzorec: C₁₃H₂₄N₂O₄
MW: 272,341 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03069732
CAS číslo: 189442-78-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR