5-Ethoxy-4-methyloxazole 95%
Dodavatel: Apollo Scientific
OR911732-1G
OR911732-5G
APOSOR911732-1GEA
1240
CZK
APOSOR911732-1G
APOSOR911732-5G
5-Ethoxy-4-methyloxazole 95%
5-Ethoxy-4-methyloxazole
Vzorec:
C₆H₉NO₂ MW: 127,141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06411139 CAS číslo: 5006-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...