Tisk…

3-Hydroxy-4-methoxycinnamic acid 95%

Dodavatel: Apollo Scientific

Synonyms: Isoferulic acid, 3-(3-Hydroxy-4-methoxyphenyl)acrylic acid

OR911490-10G OR911490-25G
APOSOR911490-10GEA 4200 CZK
APOSOR911490-10G APOSOR911490-25G
3-Hydroxy-4-methoxycinnamic acid 95%
3-Hydroxy-4-methoxycinnamic acid
Vzorec: C₁₀H₁₀O₄
MW: 194,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004391
CAS číslo: 25522-33-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR