Tisk…

tert-Butyl 4-methylbenzoate 95%

Dodavatel: Apollo Scientific

OR911439-100G OR911439-25G
APOSOR911439-100GEA 9660 CZK
APOSOR911439-100G APOSOR911439-25G
tert-Butyl 4-methylbenzoate 95%
tert-Butyl 4-methylbenzoate
Vzorec: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD06795961
CAS číslo: 13756-42-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR