Tisk…

N,N-Dimethylthiophene-3-carboxamide 97%

Dodavatel: Apollo Scientific

OR911200-1G OR911200-5G
APOSOR911200-1GEA 6220 CZK
APOSOR911200-1G APOSOR911200-5G
N,N-Dimethylthiophene-3-carboxamide 97%
N,N-Dimethylthiophene-3-carboxamide
Vzorec: C₇H₉NOS
MW: 155.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11742100
CAS číslo: 59906-37-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR