1-[(Thiophen-3-yl)carbonyl]pyrrolidine 96%
Dodavatel: Apollo Scientific
OR911199-1G
OR911199-5G
APOSOR911199-1GEA
6220
CZK
APOSOR911199-1G
APOSOR911199-5G
1-[(Thiophen-3-yl)carbonyl]pyrrolidine 96%
1-[(Thiophen-3-yl)carbonyl]pyrrolidine
Vzorec:
C₉H₁₁NOS MW: 181,255 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11742078 CAS číslo: 78909-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...