Tisk…

4-[(Thiophen-3-yl)carbonyl]morpholine 98%

Dodavatel: Apollo Scientific

OR911189-1G OR911189-5G
APOSOR911189-1GEA 5390 CZK
APOSOR911189-1G APOSOR911189-5G
4-[(Thiophen-3-yl)carbonyl]morpholine 98%
4-[(Thiophen-3-yl)carbonyl]morpholine
Vzorec: C₉H₁₁NO₂S
MW: 197,254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11742082
CAS číslo: 1090955-28-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR