Tisk…

N-Propylthiophene-3-carboxamide 98%

Dodavatel: Apollo Scientific

OR911184-1G OR911184-5G
APOSOR911184-1GEA 7670 CZK
APOSOR911184-1G APOSOR911184-5G
N-Propylthiophene-3-carboxamide 98%
N-Propylthiophene-3-carboxamide
Vzorec: C₈H₁₁NOS
Storage Temperature: Ambient
MDL Number: MFCD19595846
CAS číslo: 174359-80-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR