(3-Propoxyphenyl)acetic acid 98%
Dodavatel: Apollo Scientific
OR911179-1G
OR911179-250MG
APOSOR911179-1GEA
9330
CZK
APOSOR911179-1G
APOSOR911179-250MG
(3-Propoxyphenyl)acetic acid 98%
(3-Propoxyphenyl)acetic acid
Vzorec:
C₁₁H₁₄O₃ MW: 194.227 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07754771 CAS číslo: 900704-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...