2-[(Diethylamino)methyl]benzonitrile 98%
Dodavatel: Apollo Scientific
OR911138-10G
OR911138-1G
OR911138-5G
APOSOR911138-10GEA
16340
CZK
APOSOR911138-10G
APOSOR911138-1G
APOSOR911138-5G
2-[(Diethylamino)methyl]benzonitrile 98%
2-[(Diethylamino)methyl]benzonitrile
Vzorec:
C₁₂H₁₆N₂ MW: 188,269 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07339551 CAS číslo: 867330-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...