2-Methyl-4-nitro-1,3-benzoxazole 98%
Dodavatel: Apollo Scientific
OR911136-1G
OR911136-5G
APOSOR911136-1GEA
5200
CZK
APOSOR911136-1G
APOSOR911136-5G
2-Methyl-4-nitro-1,3-benzoxazole 98%
2-Methyl-4-nitro-1,3-benzoxazole
Vzorec:
C₈H₆N₂O₃ MW: 178,145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08064696 CAS číslo: 478553-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...