Tisk…

N-Butyl-3-nitropyridin-2-amine 98%

Dodavatel: Apollo Scientific

OR911077-1G OR911077-5G
APOSOR911077-1GEA 4770 CZK
APOSOR911077-1G APOSOR911077-5G
N-Butyl-3-nitropyridin-2-amine 98%
N-Butyl-3-nitropyridin-2-amine
Vzorec: C₉H₁₃N₃O₂
MW: 195,218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11118020
CAS číslo: 26820-68-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR