Tisk…

N-Isopropyl-3-nitropyridin-2-amine 98%

Dodavatel: Apollo Scientific

OR911064-10G OR911064-1G OR911064-5G
APOSOR911064-10GEA 21490 CZK
APOSOR911064-10G APOSOR911064-1G APOSOR911064-5G
N-Isopropyl-3-nitropyridin-2-amine 98%
N-Isopropyl-3-nitropyridin-2-amine
Vzorec: C₈H₁₁N₃O₂
MW: 181,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08692137
CAS číslo: 26820-67-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR