2-N-Ethylamino-5-nitropyridine 98%
Dodavatel: Apollo Scientific
OR911051-1G
OR911051-25G
OR911051-5G
APOSOR911051-1GEA
4140
CZK
APOSOR911051-1G
APOSOR911051-25G
APOSOR911051-5G
2-N-Ethylamino-5-nitropyridine 98%
2-N-Ethylamino-5-nitropyridine
Vzorec:
C₇H₉N₃O₂ MW: 167,165 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00023438 CAS číslo: 67730-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...