3-Methoxy-5-(4-methylphenyl)pyridine 97%
Dodavatel: Apollo Scientific
OR910874-1G
OR910874-5G
APOSOR910874-1GEA
6220
CZK
APOSOR910874-1G
APOSOR910874-5G
3-Methoxy-5-(4-methylphenyl)pyridine 97%
3-Methoxy-5-(4-methylphenyl)pyridine
Vzorec:
C₁₃H₁₃NO MW: 199,248 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22192492 CAS číslo: 1373233-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...