Tisk…

3-methoxy-5-nitrobenzonitrile 97%

Dodavatel: Apollo Scientific

OR910730-5G
APOSOR910730-5GEA 3640 CZK
APOSOR910730-5G
3-methoxy-5-nitrobenzonitrile 97%
3-methoxy-5-nitrobenzonitrile
Vzorec: C₈H₆N₂O₃
MW: 178,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463876
CAS číslo: 33224-19-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR