Tisk…

1-Methyl-6-nitroindole 98%

Dodavatel: Apollo Scientific

Synonyms: 1-Methyl-6-nitro-1H-indole

OR910612-1G OR910612-5G
APOSOR910612-1GEA 1820 CZK
APOSOR910612-1G APOSOR910612-5G
1-Methyl-6-nitroindole 98%
1-Methyl-6-nitroindole
Vzorec: C₉H₈N₂O₂
MW: 176,172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13178347
CAS číslo: 99459-48-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR