Tisk…

2-Amino-2,3-dihydroinden-1-one hydrochloride 98%

Dodavatel: Apollo Scientific

OR910599-1G OR910599-250MG
APOSOR910599-1GEA 21490 CZK
APOSOR910599-1G APOSOR910599-250MG
2-Amino-2,3-dihydroinden-1-one hydrochloride 98%
2-Amino-2,3-dihydroinden-1-one hydrochloride
Vzorec: C₉H₉NO·ClH
MW: 183,635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176559
CAS číslo: 6941-16-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR