1-Isopropyl-2-methyl-1,3-benzodiazole-5-carbonitrile 98%
Dodavatel: Apollo Scientific
OR910560-10G
OR910560-1G
OR910560-5G
APOSOR910560-10GEA
18420
CZK
APOSOR910560-10G
APOSOR910560-1G
APOSOR910560-5G
1-Isopropyl-2-methyl-1,3-benzodiazole-5-carbonitrile 98%
1-Isopropyl-2-methyl-1,3-benzodiazole-5-carbonitrile
Vzorec:
C₁₂H₁₃N₃ Storage Temperature: Ambient |
MDL Number:
MFCD20882008 CAS číslo: 1368465-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...