2-Methyl-1-propyl-1,3-benzodiazole-5-carbonitrile 98%
Dodavatel: Apollo Scientific
OR910558-1G
OR910558-25G
OR910558-5G
APOSOR910558-1GEA
4140
CZK
APOSOR910558-1G
APOSOR910558-25G
APOSOR910558-5G
2-Methyl-1-propyl-1,3-benzodiazole-5-carbonitrile 98%
2-Methyl-1-propyl-1,3-benzodiazole-5-carbonitrile
Vzorec:
C₁₂H₁₃N₃ MW: 199,252 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21467497 CAS číslo: 1403483-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...