1-Ethyl-2-methyl-1,3-benzodiazole-5-carbonitrile 98%
Dodavatel: Apollo Scientific
OR910555-1G
OR910555-25G
OR910555-5G
APOSOR910555-1GEA
4140
CZK
APOSOR910555-1G
APOSOR910555-25G
APOSOR910555-5G
1-Ethyl-2-methyl-1,3-benzodiazole-5-carbonitrile 98%
1-Ethyl-2-methyl-1,3-benzodiazole-5-carbonitrile
Vzorec:
C₁₁H₁₁N₃ MW: 185,225 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00085879 CAS číslo: 62306-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...