Tisk…

N-[(1,2-Dimethylindol-3-yl)methylidene]hydroxylamine 95%

Dodavatel: Apollo Scientific

OR910545-1G OR910545-250MG OR910545-5G
APOSOR910545-1GEA 6220 CZK
APOSOR910545-1G APOSOR910545-250MG APOSOR910545-5G
N-[(1,2-Dimethylindol-3-yl)methylidene]hydroxylamine 95%
N-[(1,2-Dimethylindol-3-yl)methylidene]hydroxylamine
Vzorec: C₁₁H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00738115
CAS číslo: 338997-02-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR