Methyl indoline-7-carboxylate 98%
Dodavatel: Apollo Scientific
OR910525-1G
OR910525-250MG
APOSOR910525-1GEA
6550
CZK
APOSOR910525-1G
APOSOR910525-250MG
Methyl indoline-7-carboxylate 98%
Methyl indoline-7-carboxylate
Vzorec:
C₁₀H₁₁NO₂ MW: 177,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09954772 CAS číslo: 112106-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...