4-Methoxycarbonyl-2,3-dihydro-1H-indole 97%
Dodavatel: Apollo Scientific
OR910523-1G
OR910523-250MG
APOSOR910523-1GEA
2360
CZK
APOSOR910523-1G
APOSOR910523-250MG
4-Methoxycarbonyl-2,3-dihydro-1H-indole 97%
4-Methoxycarbonyl-2,3-dihydro-1H-indole
Vzorec:
C₁₀H₁₁NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD07371619 CAS číslo: 155135-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...