Tisk…

4-Bromoindoline 97%

Dodavatel: Apollo Scientific

OR910522-1G OR910522-5G
APOSOR910522-1GEA 2280 CZK
APOSOR910522-1G APOSOR910522-5G
4-Bromoindoline 97%
4-Bromoindoline
Vzorec: C₈H₈BrN
MW: 198,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371633
CAS číslo: 86626-38-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR