Tisk…

7-Chloro-8-methylquinolin-2(1H)-one 95%

Dodavatel: Apollo Scientific

OR910494-100G OR910494-25G
APOSOR910494-100GEA 8340 CZK
APOSOR910494-100G APOSOR910494-25G
7-Chloro-8-methylquinolin-2(1H)-one 95%
7-Chloro-8-methylquinolin-2(1H)-one
Vzorec: C₁₀H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD15143579
CAS číslo: 73108-76-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR