Tisk…

7-Methoxyoxindole 98%

Dodavatel: Apollo Scientific

OR910474-1G OR910474-5G
APOSOR910474-1GEA 7670 CZK
APOSOR910474-1G APOSOR910474-5G
7-Methoxyoxindole 98%
7-Methoxyoxindole
Vzorec: C₉H₉NO₂
MW: 163,173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179611
CAS číslo: 7699-20-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR