Tisk…

Ethyl 1-methylpyrazole-3-carboxylate 95%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 1-methyl-1H-pyrazole-3-carboxylate

OR910459-10G OR910459-1G OR910459-5G
APOSOR910459-10GEA 12340 CZK
APOSOR910459-10G APOSOR910459-1G APOSOR910459-5G
Ethyl 1-methylpyrazole-3-carboxylate 95%
Ethyl 1-methylpyrazole-3-carboxylate
Vzorec: C₇H₁₀N₂O₂
MW: 154,166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04967314
CAS číslo: 88529-79-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR