Tisk…

Methyl 1-methylpyrrole-3-carboxylate 97%

Dodavatel: Apollo Scientific

Synonyms: Methyl 1-methyl-1H-pyrrole-3-carboxylate

OR910400-1G OR910400-250MG
APOSOR910400-1GEA 2610 CZK
APOSOR910400-1G APOSOR910400-250MG
Methyl 1-methylpyrrole-3-carboxylate 97%
Methyl 1-methylpyrrole-3-carboxylate
Vzorec: C₇H₉NO₂
MW: 139,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981621
CAS číslo: 40611-74-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR