Tisk…

(3R)-3-Ethoxypyrrolidine hydrochloride 95%

Dodavatel: Apollo Scientific

OR910377-100MG OR910377-1G OR910377-250MG
APOSOR910377-100MGEA 4570 CZK
APOSOR910377-100MG APOSOR910377-1G APOSOR910377-250MG
(3R)-3-Ethoxypyrrolidine hydrochloride 95%
(3R)-3-Ethoxypyrrolidine hydrochloride
Vzorec: C₆H₁₃NO·ClH
MW: 151.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11858548
CAS číslo: 164790-65-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR