Tisk…

7-Bromo-1-methylbenzodiazole 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 7-Bromo-1-methyl-1H-benzo[d]imidazole

OR910284-10G OR910284-1G OR910284-5G
APOSOR910284-10GEA 13530 CZK
APOSOR910284-10G APOSOR910284-1G APOSOR910284-5G
7-Bromo-1-methylbenzodiazole 98%
7-Bromo-1-methylbenzodiazole
Vzorec: C₈H₇BrN₂
MW: 211.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135915
CAS číslo: 1233542-00-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR