6-Ethoxy-1-ethylbenzimidazole 98%
Dodavatel: Apollo Scientific
OR910248-1G
OR910248-25G
OR910248-5G
APOSOR910248-1GEA
4140
CZK
APOSOR910248-1G
APOSOR910248-25G
APOSOR910248-5G
6-Ethoxy-1-ethylbenzimidazole 98%
6-Ethoxy-1-ethylbenzimidazole
Vzorec:
C₁₁H₁₄N₂O MW: 190,242 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19237196 CAS číslo: 1311197-84-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...