2-(Ethoxymethoxy)methylfuran 98%
Dodavatel: Apollo Scientific
OR910245-1G
OR910245-5G
APOSOR910245-1GEA
4770
CZK
APOSOR910245-1G
APOSOR910245-5G
2-(Ethoxymethoxy)methylfuran 98%
2-(Ethoxymethoxy)methylfuran
Vzorec:
C₈H₁₂O₃ MW: 156,179 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18837613 CAS číslo: 90199-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...