Tisk…

1-Isobutyl-1H-pyrazole 98%

Dodavatel: Apollo Scientific

OR910241-1G OR910241-5G
APOSOR910241-1GEA 6050 CZK
APOSOR910241-1G APOSOR910241-5G
1-Isobutyl-1H-pyrazole 98%
1-Isobutyl-1H-pyrazole
Vzorec: C₇H₁₂N₂
MW: 124,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08558977
CAS číslo: 725746-81-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR