Tisk…

tert-Butyl 3-methylpyrazole-1-carboxylate 95%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 3-methyl-1H-pyrazole-1-carboxylate

OR910227-1G OR910227-5G
APOSOR910227-1GEA 2860 CZK
APOSOR910227-1G APOSOR910227-5G
tert-Butyl 3-methylpyrazole-1-carboxylate 95%
tert-Butyl 3-methylpyrazole-1-carboxylate
Vzorec: C₉H₁₄N₂O₂
MW: 182,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21609566
CAS číslo: 186551-70-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR