1-(4-Methoxyphenyl)-3-methyl-1H-pyrazole 98%
Dodavatel: Apollo Scientific
OR910204-1G
OR910204-5G
APOSOR910204-1GEA
9330
CZK
APOSOR910204-1G
APOSOR910204-5G
1-(4-Methoxyphenyl)-3-methyl-1H-pyrazole 98%
1-(4-Methoxyphenyl)-3-methyl-1H-pyrazole
Vzorec:
C₁₁H₁₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD15524608 CAS číslo: 68338-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...