1-tert-Butyl-1H-pyrazole 98%
Dodavatel: Apollo Scientific
Synonyms:
1-(tert-Butyl)-1H-pyrazole
OR910151-25G
OR910151-5G
APOSOR910151-25GEA
13000
CZK
APOSOR910151-25G
APOSOR910151-5G
1-tert-Butyl-1H-pyrazole 98%
1-tert-Butyl-1H-pyrazole
Vzorec:
C₇H₁₂N₂ MW: 124,184 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12192641 CAS číslo: 15754-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...