Tisk…

1-(2-Chlorophenyl)pyrazole 98%

Dodavatel: Apollo Scientific

OR910039-1G OR910039-5G
APOSOR910039-1GEA 5790 CZK
APOSOR910039-1G APOSOR910039-5G
1-(2-Chlorophenyl)pyrazole 98%
1-(2-Chlorophenyl)pyrazole
Vzorec: C₉H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD12192626
CAS číslo: 917508-36-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR